

Chloroform ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH049-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Acetone and Aldehyde:Passes test
- Water by Karl Fischer Titration:0.02 % max.
- Acid and Chloride:Passes test
- Color (APHA):10 max.
- Free Chlorine:Passes test
- GC Analysis:99.9 % min.
- Lead:0.05 ppm max.
- Refractive Index (20ºC):1.4454 - 1.4464
- Residue:3 mg/L max.
- UV Cutoff:245 nm max.
Specification Test Results
About this item
:
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:62 °C
Melting Pt:–64 °C
Density:1,49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
:Stabilised with amylene
REACH:05-2114614906-45
:GHS07
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826