

2-Propanol:Water solution (70:30)% (v/v) stabilised, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
:67-63-0
Merck Index:12,09351
Boiling Pt:87,7 °C
Melting Pt:–62,1 °C
Density:0,785 g/cm³ (25 °C) (pure substance)
Flash Pt:-21,5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:1219
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm
REACH:05-2114615188-48
:GHS02
:GHS07
Specification Test Results
- Color (APHA):10 max.
- Isopropyl Alcohol (v/v):69 - 71 %
- Residue:1 mg/L max.
- Water (v/v):29 - 31%