

Chloroform ≥99.9% stabilised, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
:67-66-3
:
:CHCl₃
:MFCD00000826
:119.38 g/mol
:13,02160
:62 °C
:–64 °C
:1,49 g/cm³ (20 °C)
: Does not flash
:Ambient
:1888
:6.1,III
:Stabilised with amylene
:200-663-8
:05-2114614906-45
:GHS08
:GHS07
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- Electron Capture GC:10 ng/L max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4454 - 1.4464
- Residue:1 mg/L max.
- Substances Darkened by Sulfuric Acid:Passes test
- UV Cutoff:245 nm max.
- Water by Karl Fischer Titration:0.02 % max.