Order Entry
Singapore
ContactUsLinkComponent
5-Chloro-2-(trifluoromethyl)benzyl bromide ≥97%
5-Chloro-2-(trifluoromethyl)benzyl bromide ≥97%
Catalog # ALFAL19152.06
CAS Number:  
5-Chloro-2-(trifluoromethyl)benzyl bromide ≥97%
Catalog # ALFAL19152.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:261763-24-0
 :
Formula:C₈H₅BrClF₃
MDL Number:MFCD01631356
 :273.48
Boiling Pt:232 °C (1013 hPa)
Melting Pt:52 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05