Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(5-Bromopentyl)phthalimide
N-(5-Bromopentyl)phthalimide ≥97%
N-(5-Bromopentyl)phthalimide ≥97%
N-(5-Bromopentyl)phthalimide ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
N-(5-Bromopentyl)phthalimide ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
954-81-4
:
Formula:
C₁₃H₁₄BrNO₂
MDL Number:
MFCD00060522
:
296.16 g/mol
Melting Pt:
58…62 °C
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
msds_ALFAL17543
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation