1-Adamantaneethanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6240-11-5
:
Formula:C₁₂H₂₀O
MDL Number:MFCD00074756
:180.29
Boiling Pt:270 °C (1013 hPa)
Melting Pt:73…75 °C
Storage Temperature:Ambient
EINECS:228-353-8