$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
5-Bromo-o-anisidine
5-Bromo-o-anisidine ≥97%
5-Bromo-o-anisidine ≥97%
:
Thermo Fisher Scientific
5-Bromo-o-anisidine ≥97%
:
Thermo Fisher Scientific
:
6358-77-6
:
:
C₇H₈BrNO
:
MFCD04037882
:
202.05 g/mol
:
94…96 °C
:
2811
:
6.1,III
:
GHS06
:
GHS07
:
Ambient
msds_ALFAB25693
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years