Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloroethyl ethyl ether
2-Chloroethyl ethyl ether ≥98%
2-Chloroethyl ethyl ether ≥98%
:
Thermo Fisher Scientific
2-Chloroethyl ethyl ether ≥98%
:
Thermo Fisher Scientific
:
628-34-2
:
:
C₄H₉ClO
:
MFCD00000974
:
108.57 g/mol
:
107…108 °C
:
–60 °C
:
0,99
:
6 °C (43 °F)
:
Ambient
:
211-037-9
:
3271
:
3,II
:
GHS02
:
GHS07
msds_ALFAB24308
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years