2,4,5-Triphenylimidazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Lophine, 2,4,5-Triphenyl-1H-imidazole
CAS Number:484-47-9
:
Formula:C₂₁H₁₆N₂
MDL Number:MFCD00005187
Boiling Pt:509
Molecular Weight:296.37 g/mol
Melting Pt:274…278 °C
Storage Temperature:Ambient
EINECS:207-606-6
Specifications
- Environmentally Preferable: