Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethyl-4-methylimidazole
2-Ethyl-4-methylimidazole 96%
2-Ethyl-4-methylimidazole 96%
:
Thermo Fisher Scientific
2-Ethyl-4-methylimidazole 96%
:
Thermo Fisher Scientific
:
931-36-2
:
:
C₆H₁₀N₂
:
MFCD00005193
:
110.16 g/mol
:
292…295 °C
:
45…49 °C
:
0,973
:
137 °C (278 °F)
:
Ambient
:
213-234-5
:
GHS05
:
GHS07
msds_ALFAA15798
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years