


2-Methylbenzoxazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-21-6
:
Formula:C₈H₇NO
MDL Number:MFCD00005771
:133.15 g/mol
Boiling Pt:200…201 °C
Melting Pt:2…4 °C
Density:1,120
Flash Pt:85 °C (185 °F)
EINECS:202-399-9
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient