
Tris(2-methoxyphenyl)phosphine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4731-65-1
:
Formula:C₂₁H₂₁O₃P
MDL Number:MFCD00014892
:352.37
Melting Pt:196…204 °C
EINECS:225-235-8
:GHS07
Storage Temperature:Ambient