
Ethanol denatured ≥70% (v/v), Technical Grade (denatured with 1% MEK)
APPCA0565.9010
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
:
:46.07 g/mol
:Ambient
:200-578-6
:1170
:3,III
:GHS02
:64-17-5
:H₃CCH₂OH
:MFCD00003568