Order Entry
Singapore
Orders LinkContactUsLinkComponent
Hydrated PolyProbe Sampling Bag with Sampling Sponge
Hydrated PolyProbe Sampling Bag with Sampling Sponge
Hydrated polyprobe sampling bag with sampling sponge, sterile,, 8 L
Catalog # NASCB01592
Supplier:   NASCO FORT ATKINSON
Hydrated PolyProbe Sampling Bag with Sampling Sponge
Catalog # NASCB01592
Hydrated PolyProbe Sampling Bag with Sampling Sponge
Hydrated polyprobe sampling bag with sampling sponge, sterile,, 8 L
Price
$3,930.00
Log in for availability
1 * 100 items
Product Details & Documents
Make environmental surface sampling faster and easier Write-on label makes identifying samples easy and convenient Each bag contains a polyurethane sponge, premoistened with HiCap neutralizing broth Terminally gamma processed to guarantee sterility -The PolyProbe is sealed in a 24 oz. (710 ml) write-on Whirl-Pak bag and hydrated with 10 ml of HiCap Neutralizing Broth.
ags are Collection broths play a critical role in the success of an environmental monitoring program. HiCap neutralizing broth is the first collection broth developed specifically for use in the food and beverage industry as an alternative to traditional collection broths. The polyurethane sponges included with each bag are nontoxic and do not contain biocides. Unlike the wet cellulose manufacturing process, there is no need to include biocides to control microbial growth during the polymerization process.
Ideal for collecting environmental surface samples without touching the sponge. The bendable polypropylene handle allows maximum contact of the sponge with the sampling surface and also helps with the collection of hard-to-reach surfaces, around corners, and irregular shaped areas. The innovative twist-off head holds the polyurethane sponge securely but is quick and easy to remove from the handle once sample collection is completed. This listing is for of 100
Specifications
  • Capacity:
    8 L
  • Description:
    Hydrated polyprobe sampling bag with sampling sponge, sterile,