VWR® Set of 5 Single Slits
VWRMNP1000846
- Description:Set of 5 single slits
- Pk:1
Five single slits of different widths in a slide frame.
- Slit widths: 0,1/ 0,2/ 0,4/ 0,8/ 1,6 mm
- Dimensions: 50×50 mm²
Five single slits of different widths in a slide frame.