Order Entry
Singapore
ContactUsLinkComponent
Attached Lid Containers
Attached Lid Containers
  AKRO39280
 :  AKRO MILS
undefined
Attached Lid Containers
  AKRO39280
 :  AKRO MILS

 

  • Capacity:
    105,99 L
  • Colour:
    Grey
  • Material:
    Plastic
  • Seq:
    02
  • W×D×H:
    External: 457,2×635×355,6 mm
    Internal: 533,4×711,2×191,77 mm
  • Pk:
    1

 

 

Attached Lid Containers (ALCs) are reusable storage containers ideal for use in order picking, closed-loop distribution, and storage applications. Attached lids snap securely closed to protect contents from dust or damage. Comes with handle.

  • Reinforced hinge pins for years of smooth lid operation
  • Textured bottoms provide a sure grip
  • Made in the USA

These industrial shipping containers stack for maximised storage and nest when empty, saving space.

Textured bottoms provide a sure grip on conveyor belts. Strong molded-in handle grips are ergonomically designed for easy lifting and carrying. Padlock eye provides security option. Reinforced steel hinge pins ensure years of smooth lid operation.