- Thread:GL 45
- Description:ChromCap™ 100 HPLC cap assembly, adapter with vent hole and tubing, 3 ports (1,58 or 3,17 mm)
- Material:PTFE
- Pk:1
ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry.
- Patented VersaCap® technologyprovides the flexibility of interchangeable connectors
- To prevent spills and vapors fromleaking, tube fittings secure tubes tocontainer to create a close-loopsystem
- Plugs included for all unused ports
Foxx Life Sciences ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry. This system comes with patented VersaCap®, a Class VI PTFE manifold, chemically resistant viton o-ring. The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap and eliminates the tubes from sliding in and out of the container.
Each assembly comes with either a 38 to 430 mm cap or a patented GL 45 cap and manifold assembly, high quality fittings for 1×3,2 mm Ext.Ø tubing, 1×1,6 mm Ext.Ø tubing and 1×plugs of 6,4 mm for blocking of unused ports.