Order Entry
Singapore
ContactUsLinkComponent
Accessories for 1300 Series Class II, Type A2 Biological Safety Cabinets
Accessories for 1300 Series Class II, Type A2 Biological Safety Cabinets
Catalog # HERA1911322
Supplier:  Thermo Scientific
undefined
Accessories for 1300 Series Class II, Type A2 Biological Safety Cabinets
Catalog # HERA1911322
Supplier:  Thermo Scientific
Specifications
  • Description:
    Rolling caster stand for 1219,2 mm biosafety cabinet, 762 - 939,8 mm height
  • Pk:
    1
Specifications
About this item
Stands - Rolling caster stand provides comfortable fixed worksurface height of 34 inches (86 cm) for seated applications; designed for limited cabinet mobility for cleaning purposes.
Certifications: NSF/ANSI 49, Class II Type A2.