Screw Cap Decapper, 8-Channel
NUNC4105NUN
- Description:8-channel handheld screw cap capper/decapper with stand, minimum 20 racks, speed: 4 seconds to cap/decap 8 tubes, Hand operation via toggle-switch
- W×D×H:251×95×50 mm
- For use with:Nunc-style tubes
- Electrical supply:110/240 V, 50/60 Hz
- Pk:1
Process a full rack of Thermo Scientific™ Matrix™ or Thermo Scientific™ Nunc™ tubes in less than one minute with this ergonomic, manual capper/decapper. The Thermo Scientific™ 8-Channel screw cap capper/decapper is designed to cap tubes to the optimal torque to maintain sample integrity and prevent sample loss during storage. The 8-channel capper/decapper is a practical addition to benchtop processing of screw-top storage tubes.
: Warranty: one year.