Order Entry
Singapore
ContactUsLinkComponent
 
N-Methyl-2-pyrrolidone (NMP) ≥99.80% for gas chromatography, Rankem®
  RANKM0021
 :  RANKEM
 :  
N-Methyl-2-pyrrolidone (NMP) ≥99.80% for gas chromatography, Rankem®
  RANKM0021
 :  RANKEM
 :  

 

  • Pack type:
    Amber colored glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

 

 

  • Test:
    Specification
  • Guarantee Analysis:
    .
  • Assay(GC):
    ≥ 99.8 %
  • Color (APHA):
    ≤ 50
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue On Evaporation:
    ≤ 0.0005 %
  • Water (By KF):
    ≤ 0.05 %

 

 

 : 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
 :
 :99.13 g/mol
 :202 °C (1013 hPa)
 :–24 °C
 :1,033 g/cm³ (20 °C)
 :91 °C
 :212-828-1
 :01-2119472430-46
 :GHS07
 :GHS08
 :Ambient
 :872-50-4
 :C₅H₉NO
 :MFCD00003193