Ultra-Low Temperature Chambers, −40 to −80 °C, Farrar Scientific
Supplier: TRANE TECHNOLOGIES
Certificates
About this item
Forced air circulation ULT freezers specifically designed for pharmaceutical, biorepository/biobank.
- Largest volume space
- Forced-air circulation provides fastest temperature recovery from door opening
- Fully redundant systems offer peace of mind
- Small foot print
- Low temperature deviation (±1 ˚C)
- 288000 vial storage space
Designed to balance energy and have a dynamic refrigeration control.
Low energy consumption 0.33 kWh/cu. ft.
Chamber volume: 190 cu. ft./5380 L
Alarm system for high/low temperature, door ajar, power failure, remote alarm contacts.
Ordering information: Two year warranty.
Specifications
- Catalog No:
- FARR6700617W
- FARR6700617A
- Model:
- ULCI-80C-W
- ULCI-80C
- Display:
- LCD touch screen
- LCD touch screen
- Electrical:
- 230 VAC, 50 Hz, 2×30 A
- 230 VAC, 50 Hz, 2×30 A
- Weight:
- 884 kg (1950 lbs.)
- 884 kg (1950 lbs.)
- Shipping weight:
- 1184 kg (2610 lb.)
- 1184 kg (2610 lb.)