Order Entry
Singapore
ContactUsLinkComponent
Silicon dioxide amorphous, crystalline 325 Mesh
Silicon dioxide amorphous, crystalline 325 Mesh
  SPCMSI108-500GM
 :  
Silicon dioxide amorphous, crystalline 325 Mesh
  SPCMSI108-500GM
 :  
  • Pack type:
    Poly pail
  • Pk:
    500 g
  • Seq:
    3645
  • Particle size:
    325 Mesh
 :
 :60.08 g/mol
 :2230 °C (1013 hPa)
 :1427 °C
 :2,622…2,645 g/cm³ (20 °C)
 :Ambient
 :238-878-4
 :GHS08
 :7631-86-9
 :14808-60-7
 :SiO₂
 :MFCD00011232