VaporTrap™ Filter
VaporTrap™ Filter
Catalog # JUSU29916
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
Filters reduces harmful VOC vapours found inside safety cabinets.
- Cylindrical design allows 360° air flow for maximum surface contact
- High strength magnets offer flexible placement anywhere within cabinet
Improve your work environment by reducing employee's inhalation exposure to harmful VOC's. They lower emissions of VOCs into the atmosphere.
Stainless steel wire mesh cartridge containing activated carbon adsorbs volatile organic chemical vapours.
Ordering information: Two filters per package.
Specifications
- Description:VaporTrap™ Filter