Bottle Jacket
Bottle jacket for PE bottles
Catalog # JUSU56610-100
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
Bottle jacket gives the added protection of a polyethylene bottle without contaminating high purity solvents
Fits 4 L and 1 gal. bottles. Lab bottles fit to the bottom half of the cover, and lid twists securely for safe pouring. Jacket also acts as a shock absorber, minimizing breakage and spills due to bumps or falls.
Specifications
- Description:Bottle jacket for PE bottles