EDTA disodium salt 0.1 mol/l in aqueous solution titrant
Catalog # RICCR2730000-1A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037