105° vacuum connecting adapters with [ST] joints, PYREX®
Supplier: Corning
Certificates
About this item
This distillation adapter tube with sidearm has an outer joint at the top and an inner-sealed through drip tip joint at the bottom.
- 105° angle
- Top [ST] outer joint
- Bottom drip tip [ST] joint
- Serrated sidearm
Adapter tube features a [ST] outer joint at the top and an inner-sealed through drip tip [ST] joint at the bottom. The serrated sidearm is 10 mm OD.