Funnel, fluted, 60° angle, long stem, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® Funnel, Plain, 60° Angle, Long Stem, Corning
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°. These funnels are fabricated from blanks formed to a 60° angle, which permits accurate fitting of filter paper and thus reducing filtering time. In addition, few funnels have depressed inside flutings which further decrease filtering time.