Order Entry
Singapore
ContactUsLinkComponent
Fernbach-style culture flask with baffles 2800 ml, PYREX®
Fernbach-style culture flask with baffles 2800 ml, PYREX®
Catalog # CORN4423
Supplier:  Corning
undefined
Fernbach-style culture flask with baffles 2800 ml, PYREX®
Catalog # CORN4423
Supplier:  Corning

Specifications

  • Packed:
    96 tubes/rack
  • Capacity:
    1,2 ml
  • Description:
    Individual tubes, sterile
  • Seq:
    06
  • Pk:
    4.800

Specifications

About this item

This 2800 ml wide mouth PYREX® Fernbach-style culture flask is designed for culturing organisms requiring a large surface area to volume ratio. It can also be used in serum production. Triple baffled on the bottom outside edges to achieve maximal oxygen transfer to culture medium. Beaded neck rim is approximately 60 mm I.D. and accepts cotton plugs or No. 13 rubber stoppers. This flask has a large marking spot.

Caution: NOTE: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.