Order Entry
Singapore
ContactUsLinkComponent
Liquid viewer, dimmable, Apollo II
Liquid viewer, dimmable, Apollo II
  ADLM6031515
undefined
Liquid viewer, dimmable, Apollo II
  ADLM6031515
  • Description:
    Liquid viewer, dimmable, Apollo II, light level between 1,200 and 3,500 Lux
  • Pk:
    1
Designed for use in pharmaceutical and laboratory environments, the Apollo II Dimmable Liquid Viewer provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
  • Light level between 1,200 and 3,500 Lux: adjustable by dimmer control, depending on the light in the room
  • Height and angle adjustable
  • Intuitive assembly; no tools required
The Apollo II Dimmable Liquid Viewer can quickly and easily be folded for storage, with the inspection surfaces folding together to reduce chance of damage. It is packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.
Adelphi Manufacturing also offers the Apollo II High Intensity Liquid Viewer (EU & US pharmacopoeia compliant) for coloured glass containers, and the Apollo I Liquid Viewer, with additional 1.7x magnifier.
 : Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product. Supporting IQ/OQ documentation can be supplied. The user manual is available in English, with additional languages also available upon request.