Order Entry
Singapore
ContactUsLinkComponent
Accessory Kit for Digital Display Hotplates and Hotplate Stirrers, Corning®
Accessory Kit for Digital Display Hotplates and Hotplate Stirrers, Corning®
Catalog # CORN6795KIT
Supplier:  Corning
   Low Manufacturing Impact
undefined
Accessory Kit for Digital Display Hotplates and Hotplate Stirrers, Corning®
Catalog # CORN6795KIT
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Description:
    Accessory kit for digital hotplates and hotplate stirrers
  • Seq:
    08
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    1 KIT
  • For:
    PC-400D, PC-420D, PC-600D, PC-620D
Specifications
About this item
This accessory kit is designed for use with all Corning® digital display hotplates and stirring hotplates (Models PC-400D, PC-420D, PC-600D, PC-620D)
It is voltage independent. Each kit includes a temperature controller that controls the temperature of liquids up to 200 °C, a two-piece, 0.8 cm diameter, 45.7 cm long support rod, and magnetic stir bar retriever.