
Citric acid 10% (w/v), from USP materials
RICCR2120000-4A
:
- Pack type:Plastic bottle
- Pk:4 L
- Seq:5162
- Appearance:Colorless liquid
- Assay (vs. Sodium Hydroxide/Phenolphthalein):9.97-10.03 % (w/v)
:
:192.13 g/mol
:GHS05
:Ambient
:77-92-9
:HOC(COOH)(CH₂COOH)₂
:MFCD00011669