



Ammonium heptamolybdate USP Test Solution (TS)
Catalog # RICCR0674000-120B
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Poly bottle, amber
- Pk:120 mL
- Seq:2470
Specifications
Specification Test Results
- Appearance:Clear to hazy, colorless to light green with slight nitric acid odor
- Suitability for Use:Yellow precipitate with Phosphate
Specification Test Results
About this item
:
:1235.86 g/mol
Boiling Pt:~ 100 °C
Density:1,7000 g/cm³
Storage Temperature:Ambient
UN:3264
ADR:8,II
:GHS08
:GHS05
:GHS09
:GHS06
CAS Number:12054-85-2
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059