Order Entry
Singapore
ContactUsLinkComponent
Sodium carbonate 10% (w/v) in aqueous solution
Sodium carbonate 10% (w/v) in aqueous solution
Catalog # RICCR7160000-4A
Supplier:  Ricca Chemical
CAS Number:  
undefined
Sodium carbonate 10% (w/v) in aqueous solution
Catalog # RICCR7160000-4A
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay:
    10.0 ± 0.3 % (w/v)

Specification Test Results

About this item

 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494