Florisil SPE columns
Supplier: Honeywell Chemicals
Certificates
About this item
These SPE columns are made from medical-grade polypropylene with PE frits.
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- Reliable quality
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar.
Specifications
- Catalog No:
- HONC9102
- HONC9111
- HONC9104
- HONC9100
- HONC9106
- Storage:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient