Polar silica-based SPE columns
Supplier: Honeywell Chemicals
Certificates
About this item
These SPE columns are made from medical-grade polypropylene with PE frits.
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- Reliable quality
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar.
Specifications
- Catalog No:
- HONC9056
- HONC9052
- HONC9059
- HONC9062
- HONC9049
- HONC9050
- HONC9070
- HONC9073
- HONC9054
- HONC9074
- HONC9064
- HONC9068
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g