DL-α-Tocopherol acetate FCC
Catalog # SPCMV1048-100ML
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Assay:96% Min.
Specification Test Results
About this item
:
:472.75 g/mol
Density:0,96
Storage Temperature:Ambient
EINECS:231-710-0
CAS Number:7695-91-2
:52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042