Thiamine nitrate ≥98.0% FCC
SPCMT1054-100GM
:
- Pack type:Glass, amber
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
- Assay (Dried Basis):98.0 - 102.0 %
- pH of a 1 in 50 Solution:6.0 - 7.5
- Chloride (Cl):max. 0.06 %
- Lead (Pb):max. 2 mg/kg
- Loss On Drying:max. 1.0 %
: Vitamin B1 Nitrate
:
:327.36 g/mol
:Ambient
:532-43-4
:C₁₂H₁₇N₅O₄S
:MFCD00036330