Order Entry
Singapore
ContactUsLinkComponent
Nylon microtube rack
Nylon microtube rack
Catalog # HEATHS120241
Supplier:  HEATHROW
undefined
Nylon microtube rack
Catalog # HEATHS120241
Supplier:  HEATHROW

Specifications

  • For tubes:
    1,5/2,0 ml
  • Colour:
    Blue
  • Description:
    24-well nylon microtube rack
  • W×D×H:
    143L×90W×52H mm (5,6×3,5×2") mm
  • Pk:
    8

Specifications

About this item

The 24-Well Microtube rack is made of reinforced nylon, and offers superior chemical and heat resistance.

All racks are rated for use in temperatures ranging from –40 to +140 °C (–40 to +284 °F).

Integrated handles enable easy and safe transportation. Racks are stackable and available in two formats for small or large requirements. Autoclavable.