Phosphate free anionic and non-ionic detergents, Liqui-nox®
ELMI60518
- Capacity:4×1 gal.
- Description:Liqui-nox® phosphate free anionic and non-ionic detergent
- Environmentally Preferable:
- Pack type:Bottle
- Pk:4
Phosphate free anionic and non-ionic liquid detergents for manual and ultrasonic cleaning with mild pH.