384 well clear V-bottom 240 µl, polypropylene, Axygen®
AXYGP-384-240SQ-CS
: Corning
- Packed:5/pack, 50/case
- Sterile:+
- Colour:Clear
- Description:384 well V-bottom, square
- Pk:50
- Well volume:240 µl
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80°C.
- Excellent chemical resistance
- ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
- Centrifugation to 4,000 RCF
- Diamond shaped wells allow complete sample recovery
- Black and white plates are available by special order
- Ultra-flat surface ensures proper sealing with heat sealing film