Microplate, NBS, 384-well, low flange, black flat bottom, polystyrene, Corning®
CORN3654
: Corning
- No. of wells:384
- Recommended working volume:20 - 80 µl
- Sterile:-
- Colour:Black
- Description:NBS microplates
- Pk:100
- Total volume:112 µl
Black wall plates have reduced autofluorescence, higher signal to background ratio and lower CV's.
- Without lid
- Non sterile
Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions.
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise.