Order Entry
Singapore
Orders LinkContactUsLinkComponent
Media bottles, graduated, PYREX®
Media bottles, graduated, PYREX®
150 ml (5,3 oz.), Graduation interval, 25 ml; graduation range: 62 - 110 ml, GL45
Catalog # CORN1395-150
Supplier:   Corning
Media bottles, graduated, PYREX®
Catalog # CORN1395-150
Supplier:  Corning
Media bottles, graduated, PYREX®
Media bottles, graduated, PYREX®
150 ml (5,3 oz.), Graduation interval, 25 ml; graduation range: 62 - 110 ml, GL45
Price
$288.00
Log in for availability
1 * 10 items
Product Details & Documents
These PYREX® bottles are heavy-duty, reusable storage containers for media and other biological solutions.
  • Low-expansion borosilicate glass provides greater thermal strength
  • All parts withstand repeated autoclaving
Bottles have permanent white enamel graduations and a marking spot. Sizes 100 ml and above feature a glass bead that indicates full capacity line. Bottles are also ideal for dry heat sterilization (180 °C) when used with optional red PBT caps and red ETFE pouring rings.
Ordering information: Each bottle includes a linerless, one-piece, autoclavable orange polypropylene plug seal cap. All sizes except the 25 ml bottle (16157-071) include a drip-free pouring ring.
Caution: Do not tighten cap immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Bottles larger than 2 L should not be used with bottle top filter units or in other applications involving vacuum pressure as breakage may occur. A vented cap is not recommended for use on the 5 and 10 L bottles for venting during autoclaving.
Documents
Specifications
  • Cap Size:
    GL45
  • Capacity:
    150 ml (5,3 oz.)
  • Description:
    Graduation interval, 25 ml; graduation range: 62 - 110 ml