Order Entry
Singapore
ContactUsLinkComponent
 
Weighing bottle with external [ST] style opening, PYREX®
  CORN1686-3060
 :  Corning
undefined
Weighing bottle with external [ST] style opening, PYREX®
  CORN1686-3060
 :  Corning

 

  • Capacity:
    30 ml
  • Description:
    PYREX® Weighing Bottle with External [ST] Style Opening, 32 g
  • Seq:
    03
  • Pk:
    18

 

 

These PYREX® standard weighing bottles have an external 29/12 Standard Taper joint style opening.

The closure style cap fits over the externally ground body.