Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DNA denaturing solution
DNA denaturing solution
DNA denaturing solution
Corrosive
DNA denaturing solution
Catalog # ALFAJ60370.K2
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
Corrosive
DNA denaturing solution
Catalog # ALFAJ60370.K2
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
UN:
3266
ADR:
8,III
:
GHS05
MDL Number:
MFCD00146642
Product Details
Documentation