Order Entry
Singapore
ContactUsLinkComponent
VWR®, Short Thread Vials and Closures for HPLC, ND9
VWR®, Short Thread Vials and Closures for HPLC, ND9
Catalog # VWRA548-2461
Supplier:  VWR Collection
undefined
VWR®, Short Thread Vials and Closures for HPLC, ND9
Catalog # VWRA548-2461
Supplier:  VWR Collection
Specifications
  • Cap included:
    Yes
  • Septum:
    White silicone/red PTFE, bonded
  • Sterile:
    -
  • Capacity:
    1,50 ml
  • Colour:
    Clear
  • Description:
    Vials, with write-on patch
  • Seq:
    01
  • Cap type:
    PP cap with center hole
  • Cap colour:
    Blue
  • Base:
    Flat bottom
  • Pk:
    100
Specifications
About this item
USP Type-1 borosillicate glass vials , PP caps with PTFE/Silicone septa, bonded/non bonded, compatible universally on all HPLC autosampleres, prefered type vials for waters and Agilent systems.
  • Special short thread cap for LC-MS and GC-MS applications
  • One component closure - bonded
  • No bleeding
  • Completely inert
Pre-cut closures are available, where only the silicone/PTFE is slitted slightly as per major instrument manufacturer's requirement. Random batch leach free tested in HPLC/MS (report can be submitted on request).
Certifications: Each pack contains a Certificate of Confirmity (COC).
Delivery: The combopacks of vials and caps contain vials and septa caps - 100 pieces each.
Packaging: Vials are supplied completely shrink-wrapped to ensure origin, purity and transport safety.