Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrochloric acid R1
Hydrochloric acid R1 250 g/L
Hydrochloric acid R1 250 g/L
:
Ricca Chemical
Hydrochloric acid R1 250 g/L
:
Ricca Chemical
:
:
1,1500 g/cm³
:
8,II
:
GHS05
:
GHS08
:
GHS07
:
GHS06
Appearance:
Clear, colorless, hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84):
250.0 ± 0.6 g/L HCl