Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Celite® 535, filter aid
Celite® 535, filter aid, GPR RECTAPUR®
Celite® 535, filter aid, GPR RECTAPUR®
:
VWR Chemicals
Celite® 535, filter aid, GPR RECTAPUR®
:
VWR Chemicals
:
68855-54-9
:
:
SiO₂
:
MFCD00132803
:
60.08 g/mol
:
1083 °C
:
2,3…2.5 g/cm³ (20 °C)
:
GHS08
:
Ambient
pH (25 °C; 10%):
8,5 - 10,0
Loss on ignition (1000 °C):
≤ 0.5%
Particle size (< 0.1 mm):
≥ 80%
sds_VWRC22554
spec_VWRC22554.290
spec_VWRC22554.360
VWR® for Drying and Filtration