EVA PAPR with 20LF2HE loose-fitting facepiece, complete system
BULLEVAPAN4
: BULLARD
- Description:EVA PAPR with 20LF2HE Loose-fitting Facepiece
- Pk:1
World Class PAPR that is ready to use directly out of the box.
- All-in-one system ready for use, including one HEPA filter
- Filters exhalated air to keep patients safe
- Assigned protection factor (APF) 2,5× greater than an N95 mask
- Improves breathing and overall comfort compared to an N95 mask
- Loose-fitting facepiece designed for use with a stethoscope
- No fit test
- HEPA filter for use against communicable disease
EVA Powered Air-Purifying Respirator (PAPR) is equipped with the latest technology for providing wearers with constant airflow in a compact, streamlined, lightweight, ergonomic form for added comfort. Complete system includes a built-in HEPA exhalation panel to protect not only the PAPR wearer, but the environment and product as well.
Two selectable flow rate (low/high speeds) options: 7 cfm (200 lpm) and 8,5 cfm (240 lpm).
Complete EVA system includes EVA1 (blower unit), EVABELT1 (comfort closed cell foam belt), EVABAT1 (battery), EVASMC (single battery charger), PA1AFI (airflow indicator), PA20LFBT (breathing tube), 20LF2LHE facepiece, and HEPA filter.
: NIOSH certified.