EVAHL PAPRs (powered air purifying respirators), complete systems
Supplier: BULLARD
Certificates
About this item
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
- Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
- Versatile respirator for particulate or chemical environments
- Lightweight, compact, well-balanced design
- Run time: 4 to 10 hours
In addition, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8,5 CFM (high speed), 1000 Assigned Protection Factor (depending upon headtop), and dual alarms for low flow and low battery. Finally, the EVAHL has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life of a brush motor PAPR, and optional genius charger for battery trending analysis.
Complete systems include blower, battery, belt, charger, headtop, breathing tube, and airflow indicator.
Certifications: NIOSH Approvals TC-21C-0922 and TC-23C-2765. ANSI/ISA-12.12.01-2007 (CSA Certificate 2510928) Class I and II, Division and Class III, Division 1 and 2 Hazardous (Classifed) Location.
Ordering information: Filters/cartridges must be ordered separately.
Specifications
- Catalog No:
- BULLEVAHLRT3TDB
- BULLEVAHLRT3T
- BULLEVAHL20TICT
- BULLEVAHL20SICT
- BULLEVAHL20LFL
- Airflow:
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- Alarm:
- 77 dBA low voltage and 77 dBA low flow
- 77 dBA low voltage and 77 dBA low flow
- 77 dBA low voltage and 77 dBA low flow
- 77 dBA low voltage and 77 dBA low flow
- 77 dBA low voltage and 77 dBA low flow
- Battery type:
- Lithium ion polymer
- Lithium ion polymer
- Lithium ion polymer
- Lithium ion polymer
- Lithium ion polymer
- Motor:
- Brushless
- Brushless
- Brushless
- Brushless
- Brushless