Order Entry
Singapore
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Zirconium
NANOCOLOR® Photometric Tube Tests, Zirconium
Catalog # MANA985001
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Zirconium
Catalog # MANA985001
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Indicator
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Zirconium 100, test no. 0-01
  • Dimensions:
    160×110×119 mm (Including packaging)
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    36
  • Weight:
    414,1 g (Including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance; MT Easy VIS; MT Easy UV
  • Tests per kit:
    20
  • Range:
    5 - 100 mg/L (Zr)

Specifications

About this item

Photometric test kit for the determination of zirconium in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) tubes. It is pre-programmed on NANOCOLOR® photometers and is also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: No

Remark: Measuring range on NANOCOLOR VIS II

Hazardous material: No

Sludge reagent set: No

Digestion set: Yes

NanOx Metal: Yes

NanOx N: No

Delivery:

Rugged box with 20 test tubes. Sufficient for 20 tests.